trans-4-(4-n-Propylcyclohexyl)cyclohexanone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-(trans-4-Propylcyclohexyl)cyclohexanone

P2108-1G P2108-5G
TCP2108-1GEA 48.98 CAD
TCP2108-1G TCP2108-5G
trans-4-(4-n-Propylcyclohexyl)cyclohexanone ≥98.0% (by GC)
trans-4-(4-n-Propylcyclohexyl)cyclohexanone
Formula: C₁₅H₂₆O
MDL Number: MFCD04038797
CAS Number: 82832-73-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR