Fingolimod hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
F1018-1G
F1018-200MG
TCF1018-1GEA
658.86
CAD
TCF1018-1G
TCF1018-200MG
Fingolimod hydrochloride ≥98.0% (by HPLC, titration analysis)
Fingolimod hydrochloride
Formula:
C₁₉H₃₄ClNO₂ Storage Temperature: Freezer |
MDL Number:
MFCD00939512 CAS Number: 162359-56-0 Merck Index: 14,04083 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...