2,3-Diamino-5-bromo-6-methylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

D4658-1G D4658-5G
TCD4658-1GEA 154.84 CAD
TCD4658-1G TCD4658-5G
2,3-Diamino-5-bromo-6-methylpyridine ≥98.0% (by GC)
2,3-Diamino-5-bromo-6-methylpyridine
Formula: C₆H₈BrN₃
MDL Number: MFCD00160167
CAS Number: 59352-90-8
EINECS: 261-714-8

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 134.0 to 138.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR