2-Chloro-6-(trichloromethyl)pyridine ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: Nitrapyrin

C0961-1G C0961-5G
TCC0961-1GEA 64.78 CAD
TCC0961-1G TCC0961-5G
2-Chloro-6-(trichloromethyl)pyridine ≥98.0% (by GC)
2-Chloro-6-(trichloromethyl)pyridine
Formula: C₆H₃Cl₄N
MW: 230.897 g/mol
Boiling Pt: 111°C
MDL Number: MFCD00072500
CAS Number: 1929-82-4
EINECS: 217-682-2
UN: 2811
ADR: 6.1 III
Merck Index: 14,06574

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 67.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR