2-[4-(Trifluoromethyl)phenyl]ethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 2-(4-Trifluoromethylphenyl)ethylamine, 4-(Trifluoromethyl)phenethylamine

T3278-1G T3278-5G
TCT3278-1GEA 105.86 CAD
TCT3278-1G TCT3278-5G
2-[4-(Trifluoromethyl)phenyl]ethylamine ≥98.0% (by GC, titration analysis)
2-[4-(Trifluoromethyl)phenyl]ethylamine
Formula: C₉H₁₀F₃N
MW: 189.181 g/mol
Boiling Pt: 90°C
Storage Temperature: Refrigerator
MDL Number: MFCD01529875
CAS Number: 775-00-8
UN: 2735
ADR: 8 II

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR