4-Formylbenzeneboronic acid pinacol ester ≥97.0% (by GC)

Supplier: TCI America

T2923-1G T2923-5G
TCT2923-1GEA 37.92 CAD
TCT2923-1G TCT2923-5G
4-Formylbenzeneboronic acid pinacol ester ≥97.0% (by GC)
4-Formylbenzeneboronic acid pinacol ester
Formula: C₁₃H₁₇BO₃
MDL Number: MFCD04972375
CAS Number: 128376-64-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 59.0 to 63.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR