N-Methylpyridin-3-amine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3-(Methylamino)pyridine

M2684-1G M2684-5G
TCM2684-1GEA 83.74 CAD
TCM2684-1G TCM2684-5G
N-Methylpyridin-3-amine ≥98.0% (by GC, titration analysis)
N-Methylpyridin-3-amine
Formula: C₆H₈N₂
Boiling Pt: 110°C
MDL Number: MFCD01418130
CAS Number: 18364-47-1

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR