Cinnarizine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

C3459-10G C3459-50G
TCC3459-10GEA 75.84 CAD
TCC3459-10G TCC3459-50G
Cinnarizine ≥98.0% (by HPLC, total nitrogen)
Cinnarizine
Formula: C₂₆H₂₈N₂
Storage Temperature: Refrigerator
MDL Number: MFCD00056037
CAS Number: 298-57-7
EINECS: 206-064-8
Merck Index: 14,02305

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR