2,2'-Bicinchoninic acid disodium salt, hydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
B4509-1G
B4509-5G
TCB4509-1GEA
39.5
CAD
TCB4509-1G
TCB4509-5G
2,2'-Bicinchoninic acid disodium salt, hydrate ≥98.0% (by HPLC, titration analysis)
2,2'-Bicinchoninic acid disodium salt
Formula:
C₂₀H₁₀N₂Na₂O₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00037500 CAS Number: 979-88-4 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Water | 3.0 to 16.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...