Cefdinir ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

C3111-1G C3111-5G
TCC3111-1GEA 167.48 CAD
TCC3111-1G TCC3111-5G
Cefdinir ≥97.0% (by HPLC, titration analysis)
Cefdinir
Formula: C₁₄H₁₃N₅O₅S₂
Melting Pt: 190°C
MDL Number: MFCD00865030
CAS Number: 91832-40-5
Merck Index: 14,01920

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Specific rotation [a]20/D -58.0 to -66.0 deg (C=1,0.1mol/L buffer sol. pH7.0 Phosphate)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR