Mupirocin ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M2955-100MG M2955-25MG
TCM2955-100MGEA 764.72 CAD
TCM2955-100MG TCM2955-25MG
Mupirocin ≥98.0% (by titrimetric analysis)
Mupirocin
Formula: C₂₆H₄₄O₉
Melting Pt: 78°C
MDL Number: MFCD01711620
CAS Number: 12650-69-0
Merck Index: 14,06302

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -18.0 to -21.0 deg (C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR