2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: N-(2-Methyl-5-nitrophenyl)-4-(3-pyridyl)-2-pyrimidinamine, 2-(2-Methyl-5-nitroanilino)-4-(3-pyridyl)pyrimidine

M2584-1G M2584-5G
TCM2584-1GEA 61.62 CAD
TCM2584-1G TCM2584-5G
2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine ≥98.0% (by HPLC, titration analysis)
2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine
Formula: C₁₆H₁₃N₅O₂
MDL Number: MFCD02179269
CAS Number: 152460-09-8

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 192.0 to 196.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR