N,N'-Di(1-naphthyl)benzidine ≥98.0% (by HPLC, total nitrogen), purified by sublimation

Supplier: TCI America

Synonyms: N,N’-Di(1-naphthyl)-4,4’-benzidine

D4768-1G D4768-5G
TCD4768-1GEA 104.28 CAD
TCD4768-1G TCD4768-5G
N,N'-Di(1-naphthyl)benzidine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
N,N'-Di(1-naphthyl)benzidine
Formula: C₃₂H₂₄N₂
MW: 436.558 g/mol
MDL Number: MFCD09261380
CAS Number: 152670-41-2

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR