L-Aspartic aciddiethyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D4756-25G D4756-5G
TCD4756-25GEA 135.88 CAD
TCD4756-25G TCD4756-5G
L-Aspartic aciddiethyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
L-Aspartic aciddiethyl ester hydrochloride
Formula: C₈H₁₅NO₄·HCl
MDL Number: MFCD00038910
CAS Number: 16115-68-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 105.0 to 109.0 deg-C
Specific rotation [a]20/D +7.0 to +9.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR