2-bromo-5-methylthiazole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B4604-1G
B4604-5G
TCB4604-1GEA
60.04
CAD
TCB4604-1G
TCB4604-5G
2-bromo-5-methylthiazole ≥98.0% (by GC)
2-bromo-5-methylthiazole
Formula:
C₄H₄BrNS Boiling Pt: 84°C |
MDL Number:
MFCD08460610 CAS Number: 41731-23-1 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...