Fmoc-N-Me-Ile-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F1129-1G F1129-5G
TCF1129-1GEA 67.94 CAD
TCF1129-1G TCF1129-5G
Fmoc-N-Me-Ile-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-N-Me-Ile-OH
Formula: C₂₂H₂₅NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00153389
CAS Number: 138775-22-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 172.0 to 177.0 deg-C
Specific rotation [a]20/D -59.0 to -64.0 deg (C=1,DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR