(5-bromothien-2-yl)methanol ≥97.0% (by GC)
Supplier: TCI America
B4651-1G
B4651-5G
TCB4651-1GEA
96.38
CAD
TCB4651-1G
TCB4651-5G
(5-bromothien-2-yl)methanol ≥97.0% (by GC)
(5-bromothien-2-yl)methanol
Formula:
C₅H₅BrOS |
MDL Number:
MFCD06202626 CAS Number: 79387-71-6 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...