N-Boc-D-serine methyl ester ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B4859-1G B4859-5G
TCB4859-1GEA 41.08 CAD
TCB4859-1G TCB4859-5G
N-Boc-D-serine methyl ester ≥98.0% (by HPLC, total nitrogen)
N-Boc-D-serine methyl ester
Formula: C₉H₁₇NO₅
MDL Number: MFCD00270516
CAS Number: 95715-85-8

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D +15.0 to +21.0 deg (C=5, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR