Ethyl (2-fluorobenzoyl)acetate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl 3-(2-fluorophenyl)-3-oxo-propionate

E1097-25G E1097-5G
TCE1097-25GEA 630.42 CAD
TCE1097-25G TCE1097-5G
Ethyl (2-fluorobenzoyl)acetate ≥98.0% (by GC)
Ethyl (2-fluorobenzoyl)acetate
Formula: C₁₁H₁₁FO₃
MW: 210.204 g/mol
Boiling Pt: 108°C
MDL Number: MFCD00000321
CAS Number: 1479-24-9

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 1.1740 to 1.1860


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR