2-Morpholinoacetic acid hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Morpholinoacetic acid hydrochloride, Morpholin-4-ylacetic acid hydrochloride, 4-Morpholineacetic acid hydrochloride
M2608-1G
M2608-5G
TCM2608-1GEA
60.04
CAD
TCM2608-1G
TCM2608-5G
2-Morpholinoacetic acid hydrochloride ≥98.0% (by titrimetric analysis)
2-Morpholinoacetic acid hydrochloride
Formula:
C₆H₁₂ClNO₃ MW: 181.616 g/mol |
MDL Number:
MFCD03540888 CAS Number: 89531-58-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...