Bis(p-tolyl)phosphine oxide ≥95.0% (by GC)

Supplier: TCI America
Warning

D4520-1G D4520-5G
TCD4520-1GEA 159.58 CAD
TCD4520-1G TCD4520-5G
Bis(p-tolyl)phosphine oxide ≥95.0% (by GC)
Bis(p-tolyl)phosphine oxide
Formula: C₁₄H₁₅OP
Melting Pt: 96°C
MDL Number: MFCD01445489
CAS Number: 2409-61-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR