2,3-Difluoro-4-n-propoxybenzeneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: (2,3-Difluoro-4-propoxyphenyl)boronic acid, 2,3-Difluoro-4-propoxyphenylboronic acid, 2,3-Difluoro-4-propoxybenzeneboronic acid

D5083-1G D5083-5G
76219-844EA 113.76 CAD
76219-844 76219-846
2,3-Difluoro-4-n-propoxybenzeneboronic acid (contains varying amounts of Anhydride)
2,3-Difluoro-4-n-propoxybenzeneboronic acid
Formula: C₉H₁₁BF₂O₃
MDL Number: MFCD14584457
CAS Number: 212837-49-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR