Ethyl-4-methylthiazole-5-carboxylate ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
4-Methylthiazole-5-carboxylic acid ethyl ester, Ethyl 4-methylthiazole-5-carboxylate
E1182-1G
E1182-5G
TCE1182-1GEA
107.44
CAD
TCE1182-1G
TCE1182-5G
Ethyl-4-methylthiazole-5-carboxylate ≥97.0% (by GC)
Ethyl-4-methylthiazole-5-carboxylate
Formula:
C₇H₉NO₂S MW: 171.214 g/mol Boiling Pt: 233°C Melting Pt: 28°C |
MDL Number:
MFCD00626873 CAS Number: 20582-55-2 |
Specification Test Results
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...