2,2':6',2''-Terpyridine-4'-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: [2,2':6',2''-Terpyridine]-4'-carboxylic acid

T3245-1G T3245-200MG
TCT3245-1GEA 405.14 CAD
TCT3245-1G TCT3245-200MG
2,2':6',2''-Terpyridine-4'-carboxylic acid ≥98.0% (by GC, titration analysis)
2,2':6',2''-Terpyridine-4'-carboxylic acid
Formula: C₁₆H₁₁N₃O₂
MW: 277.283 g/mol
MDL Number: MFCD04114290
CAS Number: 148332-36-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR