4-Hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane-1-oxide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,6,7-Trioxa-1-phosphabicyclo[2.2.2]octane-4-methanol 1-oxide

H1548-1G H1548-5G
TCH1548-1GEA 71.1 CAD
TCH1548-1G TCH1548-5G
4-Hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane-1-oxide ≥98.0% (by GC)
4-Hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane-1-oxide
Formula: C₅H₉O₅P
MW: 180.096 g/mol
MDL Number: MFCD01677612
CAS Number: 5301-78-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR