1-n-Butoxy-2,3-difluorobenzene ≥98.0% (by GC)
Supplier: TCI America
B5058-25G
B5058-5G
TCB5058-25GEA
99.22
CAD
TCB5058-25G
TCB5058-5G
1-n-Butoxy-2,3-difluorobenzene ≥98.0% (by GC)
1-n-Butoxy-2,3-difluorobenzene
Formula:
C₁₀H₁₂F₂O Boiling Pt: 110°C |
MDL Number:
MFCD07368738 CAS Number: 136239-66-2 EINECS: 446-810-1 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...