2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
M2800-1G
M2800-5G
TCM2800-1GEA
97.96
CAD
TCM2800-1G
TCM2800-5G
2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole ≥98.0% (by HPLC, titration analysis)
2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole
Formula:
C₁₆H₁₄F₃N₃OS Storage Temperature: Freezer |
MDL Number:
MFCD00834357 CAS Number: 103577-40-8 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...