Methyl-5-bromo-2-methylbenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

M2542-1G M2542-5G
TCM2542-1GEA 30.02 CAD
TCM2542-1G TCM2542-5G
Methyl-5-bromo-2-methylbenzoate ≥98.0% (by GC)
Methyl-5-bromo-2-methylbenzoate
Formula: C₉H₉BrO₂
Boiling Pt: 265°C
MDL Number: MFCD09839555
CAS Number: 79669-50-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 50.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR