4-(Methoxymethyl)benzeneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: (4-(Methoxymethyl)phenyl)boronic acid4-(Methoxymethyl)phenylboronic acid

M2677-5G M2677-1G
TCM2677-5GEA 277.1 CAD
TCM2677-5G TCM2677-1G
4-(Methoxymethyl)benzeneboronic acid (contains varying amounts of Anhydride)
4-(Methoxymethyl)benzeneboronic acid
Formula: C₈H₁₁BO₃
MW: 165.983 g/mol
MDL Number: MFCD03788426
CAS Number: 279262-11-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR