TOTT ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
T3569-25G
T3569-5G
TCT3569-25GEA
382.36
CAD
TCT3569-25G
TCT3569-5G
TOTT ≥98.0% (by HPLC)
TOTT
Formula:
C₁₀H₁₆BF₄N₃OS Storage Temperature: Freezer |
MDL Number:
MFCD05664718 CAS Number: 255825-38-8 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...