N-(Benzoyloxy)succinimide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 2,5-Dioxopyrrolidin-1-yl benzoate, Bz-OSu

B4785-25G B4785-5G
TCB4785-25GEA 535.62 CAD
TCB4785-25G TCB4785-5G
N-(Benzoyloxy)succinimide ≥98.0% (by HPLC, total nitrogen)
N-(Benzoyloxy)succinimide
Formula: C₁₁H₉NO₄
MW: 219.196 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00078953
CAS Number: 23405-15-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 135.0 to 139.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR