5-Bromo-4-chloro-3-indolyl acetate ≥98.0% (by HPLC)

Supplier: TCI America

B4830-200MG B4830-50MG
TCB4830-200MGEA 363.4 CAD
TCB4830-200MG TCB4830-50MG
5-Bromo-4-chloro-3-indolyl acetate ≥98.0% (by HPLC)
5-Bromo-4-chloro-3-indolyl acetate
Formula: C₁₀H₇BrClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00037932
CAS Number: 3252-36-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR