4-Methylphenylalanine 98,0 HPLC, T

Supplier: TCI America

Synonyms: H-Phe(4-Me)-OH, H-p-Me-Phe-OH, 4-Methyl-L-phenylalanine

M2743-1G M2743-5G
TCM2743-1GEA 159.58 CAD
TCM2743-1G TCM2743-5G
4-Methylphenylalanine 98,0 HPLC, T
4-Methylphenylalanine
Formula: C₁₀H₁₃NO₂
MW: 179.219 g/mol
MDL Number: MFCD00038575
CAS Number: 1991-87-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -8.0 to -11.0 deg (C=1,1mol/L HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR