H-D-Lys(Z)-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C3345-25G C3345-5G
TCC3345-25GEA 319.16 CAD
TCC3345-25G TCC3345-5G
H-D-Lys(Z)-OH ≥98.0% (by HPLC, titration analysis)
H-D-Lys(Z)-OH
Formula: C₁₄H₂₀N₂O₄
MDL Number: MFCD00063120
CAS Number: 34404-32-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -15.0 to -17.0 deg (C=1.6,2mol/L HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR