β-Alanine-tert-butylester hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: beta-Alanine tert-Butyl ester hydrochloride, β-Alanine t-butyl ester hydrochloride, H-β-Ala-OtBu·HCl, H-beta-Ala-OtBu·HCl, H-beta-Ala-OtBu.HCl, H-β-Ala-OtBu.HCl, tert-Butyl 3-aminopropanoate hydrochloride

A2772-25G A2772-5G
TCA2772-25GEA 292.3 CAD
TCA2772-25G TCA2772-5G
β-Alanine-tert-butylester hydrochloride ≥98.0% (by titrimetric analysis)
beta-Alanine-tert-butylester hydrochloride
Formula: C₇H₁₆ClNO₂
Storage Temperature: Freezer
MDL Number: MFCD00058258
CAS Number: 58620-93-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR