N-Acetyl-DL-phenylalanine 98,0 HPLC, T

Supplier: TCI America

Synonyms: Ac-DL-Phe-OH

A2864-25G A2864-5G
TCA2864-25GEA 64.78 CAD
TCA2864-25G TCA2864-5G
N-Acetyl-DL-phenylalanine 98,0 HPLC, T
N-Acetyl-DL-phenylalanine
Formula: C₁₁H₁₃NO₃
MW: 207.229 g/mol
MDL Number: MFCD00063157
CAS Number: 2901-75-9
EINECS: 220-793-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 152.0 to 156.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR