L-Valine ethyl ester hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: H-Val-OEt·HCl, (S)-Ethyl 2-amino-3-methylbutanoate hydrochloride

V0142-25G V0142-5G
TCV0142-25GEA 83.74 CAD
TCV0142-25G TCV0142-5G
L-Valine ethyl ester hydrochloride ≥98.0% (by titrimetric analysis)
L-Valine ethyl ester hydrochloride
Formula: C₇H₁₆ClNO₂
Melting Pt: 104°C
MDL Number: MFCD00012511
CAS Number: 17609-47-1
EINECS: 241-580-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +6.0 to +8.0 deg (C=2,H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR