Z-Glu-OtBu ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1-tert-Butyl N-Carbobenzoxy-L-glutamate, N-Cbz-L-glutamic acid 1-tert-Butyl ester, 1-tert-Butyl N-Cbz-L-glutamate, N-Carbobenzoxy-L-glutamic acid 1-tert-Butyl ester, Z-Glu-OtBu, 1-tert-Butyl N-Benzyloxycarbonyl-L-glutamate, N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester, N-Benzyloxycarbonyl-L-glutamic acid 1-tert-Butyl ester

B4912-1G B4912-5G
TCB4912-1GEA 75.84 CAD
TCB4912-1G TCB4912-5G
Z-Glu-OtBu ≥98.0% (by HPLC, titration analysis)
Z-Glu-OtBu
Formula: C₁₇H₂₃NO₆
MW: 337.372 g/mol
MDL Number: MFCD00077016
CAS Number: 5891-45-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 81.0 to 85.0 deg-C
Specific rotation [a]20/D -24.0 to -28.0 deg (C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR