Z-Aib-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-Benzyloxycarbonyl-alpha-methylalanine, 2-(Benzyloxycarbonylamino)isobutyric acid, N-Benzyloxycarbonyl-2-methylalanine, Z-Aib-OH, 2-(Cbz-amino)isobutyric acid, 2-(Carbobenzoxyamino)isobutyric acid, N-Cbz-alpha-methylalanine

C2761-5G C2761-25G
76218-888EA 58.46 CAD
76218-888 76219-684
Z-Aib-OH ≥98.0% (by HPLC, titration analysis)
Z-Aib-OH
Formula: C₁₂H₁₅NO₄
MDL Number: MFCD00004190
CAS Number: 15030-72-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 77.0 to 81.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR