Z-Pyr-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (S)-1-Carbobenzoxy-5-oxopyrrolidine-2-carboxylic acid, N-Cbz-L-pyroglutamic acid, (S)-1-Benzyloxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, Z-Pyr-OH, N-Benzyloxycarbonyl-L-pyroglutamic acid, N-Carbobenzoxy-L-pyroglutamic acid, (S)-1-Cbz-5-oxopyrrolidine-2-carboxylic acid, (S)-1-((Benzyloxy)carbonyl)-5-oxopyrrolidine-2-carboxylic acid

C3143-25G C3143-5G
TCC3143-25GEA 252.8 CAD
TCC3143-25G TCC3143-5G
Z-Pyr-OH ≥98.0% (by HPLC, titration analysis)
Z-Pyr-OH
Formula: C₁₃H₁₃NO₅
Storage Temperature: Freezer
MDL Number: MFCD00037352
CAS Number: 32159-21-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 139.0 to 143.0 deg-C
Specific rotation [a]20/D -34.0 to -37.0 deg (C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR