Fmoc-Lys(Ac)-OH ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
F1041-1G
F1041-5G
TCF1041-1GEA
94.8
CAD
TCF1041-1G
TCF1041-5G
Fmoc-Lys(Ac)-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-Lys(Ac)-OH
Formula:
C₂₃H₂₆N₂O₅ Storage Temperature: Freezer |
MDL Number:
MFCD00077409 CAS Number: 159766-56-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 162.0 to 166.0 deg-C |
Specific rotation [a]20/D | -15.0 to -17.0 deg (C=1,DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...