Fmoc-Inp-OH 98,0 HPLC, T
Supplier: TCI America
Synonyms:
1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-piperidinecarboxylic acid, 1-Fmoc-isonipecotic acid, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]isonipecotic acid, Fmoc-isonipecotic acid, 1-Fmoc-4-piperidinecarboxylic acid, Fmoc-Isn-OH, 1-Fmoc-piperidine-4-carboxylic acid
F0914-1G
F0914-5G
TCF0914-1GEA
58.46
CAD
TCF0914-1G
TCF0914-5G
Fmoc-Inp-OH 98,0 HPLC, T
Fmoc-Inp-OH
Formula:
C₂₁H₂₁NO₄ MW: 351.402 g/mol |
MDL Number:
MFCD00273475 CAS Number: 148928-15-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...