Fmoc-4-fluoro-L-phenylalanine ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

F0902-1G F0902-5G
TCF0902-1GEA 135.56 CAD
TCF0902-1G TCF0902-5G
Fmoc-4-fluoro-L-phenylalanine ≥95.0% (by HPLC, titration analysis)
Fmoc-4-fluoro-L-phenylalanine
Formula: C₂₄H₂₀FNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00191197
CAS Number: 169243-86-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 95.0 %
Melting point 182.0 to 186.0 deg-C
Specific rotation [a]20/D -36.0 to -40.0 deg (C=1,DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR