Fmoc-Glu-OAll ≥96.0% (by HPLC, titration analysis)
Supplier: TCI America
A2909-1G
A2909-5G
TCA2909-1GEA
88.48
CAD
TCA2909-1G
TCA2909-5G
Fmoc-Glu-OAll ≥96.0% (by HPLC, titration analysis)
Fmoc-Glu-OAll
Formula:
C₂₃H₂₃NO₆ Storage Temperature: Refrigerator |
MDL Number:
MFCD00467718 CAS Number: 144120-54-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Neutralization titration) | min. 96.0 % |
Melting point | 117.0 to 121.0 deg-C |
Specific rotation [a]20/D | -17.0 to -21.0 deg (C=1,DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...