Fmoc-Asp-OAll ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-Fmoc-L-aspartic acid 1-Allyl ester, Fmoc-Asp-allyl ester, N-Fmoc-L-aspartic acid 1-allyl ester, 1-Allyl N-Fmoc-L-aspartate, Fmoc-Asp-OAll, 1-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate, N-[(9H-Fluoren-9-ylmethyloxycarbonyl)]-L-aspartic acid 1-Allyl ester

A2894-1G A2894-5G
TCA2894-1GEA 134.3 CAD
TCA2894-1G TCA2894-5G
Fmoc-Asp-OAll ≥98.0% (by HPLC, titration analysis)
Fmoc-Asp-OAll
Formula: C₂₂H₂₁NO₆
MW: 395.411 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00467715
CAS Number: 144120-53-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 131.0 to 135.0 deg-C
Specific rotation [a]20/D -22.0 to -26.0 deg (C=1,DMF)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR