Fmoc-Lys(biotin)-OH ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Nα-Biotinyl-Nepsilon-Fmoc-L-lysine, Fmoc-Biocytin, (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-(5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexanoic acid, Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-biotinyl-L-lysine, Fmoc-Lys(biotinyl)-OH, Nalpha-Fmoc-Nepsilon-biotinyl-L-lysine

F1042-1G F1042-200MG
TCF1042-1GEA 360.13 CAD
TCF1042-1G TCF1042-200MG
Fmoc-Lys(biotin)-OH ≥95.0% (by HPLC, titration analysis)
Fmoc-Lys(biotin)-OH
Formula: C₃₁H₃₈N₄O₆S
MW: 594.727 g/mol
MDL Number: MFCD00270741
CAS Number: 146987-10-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 95.0 %
Specific rotation [a]20/D +18.0 to +21.0 deg (C=1,DMSO)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR