Fmoc-D-Arg(Pbf)-OH ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulphonyl)-D-arginine, Fmoc-D-Arg(Pbf)-OH, Nα-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-D-arginine, Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-D-arginine, Nα-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-D-arginine, Nalpha-Fmoc-Nomega-Pbf-D-arginine
F0875-1G
F0875-5G
TCF0875-1GEA
60.04
CAD
TCF0875-1G
TCF0875-5G
Fmoc-D-Arg(Pbf)-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-D-Arg(Pbf)-OH
Formula:
C₃₄H₄₀N₄O₇S MW: 648.775 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00237010 CAS Number: 187618-60-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +4.5 to +6.5 deg (C=1、DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...