Fmoc-Lys(Mtt)-OH 98,0 HPLC, N

Supplier: TCI America

Synonyms: Nα-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-(4-methyltrityl)-L-lysine, Nα-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine

F1043-1G F1043-5G
TCF1043-1GEA 112.18 CAD
TCF1043-1G TCF1043-5G
Fmoc-Lys(Mtt)-OH 98,0 HPLC, N
Fmoc-Lys(Mtt)-OH
Formula: C₄₁H₄₀N₂O₄
MW: 624.781 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237166
CAS Number: 167393-62-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -9.0 to -11.0 deg (C=1,DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR