Fmoc-d-pipecolic acid 98,0 HPLC, T

Supplier: TCI America
Warning

Synonyms: (R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-2-carboxylic acid, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-pipecolic acid, (R)-1-Fmoc-2-piperidinecarboxylic acid, Fmoc-D-Pip-OH, 1-Fmoc-D-pipecolic acid, Fmoc-D-Homopro-OH, (R)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid

F0937-1G F0937-5G
TCF0937-1GEA 83.74 CAD
TCF0937-1G TCF0937-5G
Fmoc-d-pipecolic acid 98,0 HPLC, T
Fmoc-d-pipecolic acid
Formula: C₂₁H₂₁NO₄
MW: 351.402 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00235899
CAS Number: 101555-63-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 155.0 to 159.0 deg-C
Specific rotation [a]20/D +21.0 to +25.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR