Fmoc-L-norvaline ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-norvaline, Fmoc-Nva-OH, N-Fmoc-L-norvaline

F1117-1G F1117-5G
TCF1117-1GEA 41.08 CAD
TCF1117-1G TCF1117-5G
Fmoc-L-norvaline ≥98.0% (by HPLC, titration analysis)
Fmoc-L-norvaline
Formula: C₂₀H₂₁NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00155631
CAS Number: 135112-28-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 153.0 to 157.0 deg-C
Specific rotation [a]20/D -20.0 to -22.0 deg (C=2, DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR